Novus Biologicals
Manufacturer Code:NBP2443440.2MG
Catalog # NBP24434402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | EMI1/1176 |
Immunogen | Recombinant fragment (203 amino acid residues between aa 1-250) of human EMI1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Early mitotic inhibitor 1; EMI1Early mitotic inhibitor 1 F-box only protein 5 F-box protein 5 FBX5F-box protein Fbx5 Fbxo31; F-box only protein 5; F-box protein Fbx5
Gene Aliases: EMI1; FBX5; Fbxo31; FBXO5
UniProt ID: (Human) Q9UKT4
Entrez Gene ID: (Human) 26271
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.