Invitrogen
Manufacturer Code:PA541812
Catalog # PIPA541812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human EMG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 18S rRNA (pseudouridine(1248)-N1)-methyltransferase; 18S rRNA (pseudouridine-N1-)-methyltransferase NEP1; 18S rRNA Psi1248 methyltransferase; EMG1 N1-specific pseudouridine methyltransferase; EMG1 nucleolar protein homolog; essential for mitotic growth 1; gene rich cluster, C2f; nucleolar essential protein 1; Nucleolar protein EMG1 homolog; probable ribosome biogenesis protein NEP1; Protein C2f; Ribosomal RNA small subunit methyltransferase NEP1; Ribosome biogenesis protein NEP1
Gene Aliases: C2F; EMG1; Grcc2f; NEP1; zgc:136360
UniProt ID: (Human) Q92979, (Mouse) O35130
Entrez Gene ID: (Human) 10436, (Horse) 100052984, (Dog) 477708, (Bovine) 515362, (Rabbit) 100342038, (Guinea pig) 100722865, (Mouse) 14791, (Rat) 312706, (Zebrafish) 678588
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.