Proteintech
Manufacturer Code:129751AP150UL
Catalog # 50-172-8389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EME1 Fusion Protein Ag3580 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Crossover junction endonuclease EME1; EME1 hMMS4 MMS4 MMS4 homolog MMS4L; essential meiotic endonuclease 1 homolog 1; essential meiotic endonuclease 1 homolog 2; hMMS4; homolog of yeast EME1 endonuclease; MMS4 homolog; SLX2 structure-specific endonuclease subunit homolog A
Gene Aliases: 6820428D13; EME1; MMS4; MMS4L; SLX2A
UniProt ID: (Human) Q96AY2, (Mouse) Q8BJW7
Entrez Gene ID: (Human) 146956, (Mouse) 268465, (Rat) 287634
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.