OriGene
Manufacturer Code:TA809145
Catalog # 50-168-2598
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4B2 |
Immunogen | Human recombit protein fragment corresponding to amino acids 133-407 of human ELP4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AN AN2 C11orf19 PAX6NEB PAXNEB dJ68P15A.1 hELP4; elongation protein 4 homolog; Elongator complex protein 4; hELP4; PAX6 neighbor gene protein
Gene Aliases: AN; C11orf19; dJ68P15A.1; ELP4; hELP4; PAX6NEB; PAXNEB
UniProt ID: (Human) Q96EB1
Entrez Gene ID: (Human) 26610
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.