Invitrogen
Manufacturer Code:PA568851
Catalog # PIPA568851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse ELP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C11orf19 PAX6NEB PAXNEB dJ68P15A.1 PAX6 neighbor gene protein elongator complex protein 4 hELP4; Elongator complex protein 4; PAX6 neighbor gene protein
Gene Aliases: A330107A17Rik; Elp4; Paxneb
UniProt ID: (Mouse) Q9ER73
Entrez Gene ID: (Mouse) 77766, (Rat) 687694
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.