Invitrogen
Manufacturer Code:PA555260
Catalog # PIPA555260
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ELP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: elongation protein 3 homolog; Elongator complex protein 3; Elongator complex protein 3 EC 2.3.1.48 hELP3 ELP3; hELP3; Protein lysine acetyltransferase ELP3; rCG52086-like; tRNA uridine(34) acetyltransferase
Gene Aliases: 2610507P14Rik; ELP3; KAT9
UniProt ID: (Human) Q9H9T3, (Mouse) Q9CZX0
Entrez Gene ID: (Human) 55140, (Mouse) 74195, (Rat) 100362548
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.