Invitrogen
Manufacturer Code:PA5103827
Catalog # PIPA5103827
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ELP3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610507P14Rik EC 2.3.1.48 elongation protein 3 homolog elongator acetyltransferase complex subunit 3 elongator complex protein 3 Elp3 h KAT9 hELP3 KAT9 LOC100362548 Protein lysine acetyltransferase ELP3 rCG52086-like tRNA uridine(34) acetyltransferase; elongation protein 3 homolog; Elongator complex protein 3; hELP3; Protein lysine acetyltransferase ELP3; tRNA uridine(34) acetyltransferase
Gene Aliases: 2610507P14Rik; ELP3; KAT9
UniProt ID: (Human) Q9H9T3, (Mouse) Q9CZX0
Entrez Gene ID: (Human) 55140, (Mouse) 74195
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.