Invitrogen
Manufacturer Code:PA531859
Catalog # PIPA531859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 318 and 620 of Human ELP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: elongation protein 2 homolog; Elongator complex protein 2; elongator protein 2; ELP2; SHINC-2; SHINC-2 STATIP1 StIP STAT3-interacting protein 1 elongator complex protein 2 elongator protein 2 signal transducer and activator of transcription 3 interacting protein 1 signal transducer and activator of transcription interacting protein 1 stIP1; signal transducer and activator of transcription 3 interacting protein 1; STAT3-interacting protein 1; StIP1
Gene Aliases: ELP2; SHINC-2; STATIP1; StIP
UniProt ID: (Human) Q6IA86
Entrez Gene ID: (Human) 55250
Molecular Function: RNA binding protein enzyme modulator esterase hydrolase kinase inhibitor kinase modulator mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.