Novus Biologicals
Manufacturer Code:NBP250384V
Catalog # NBP250384V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | Z26 |
Immunogen | Peptide Sequence - RAYTKGQRLP (peptide immunogen is identical in rat (Rattus norvegicus) mouse (Mus musculus) cow (Bos taurus) dog (Canis familiaris) horse (Equus caballus) |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3-keto acyl-CoA synthase ELOVL7; Elongation of very long chain fatty acids protein 7; ELOVL FA elongase 7; ELOVL family member 7 elongation of long chain fatty acids ELOVL fatty acid elongase 7; ELOVL family member 7, elongation of long chain fatty acids; ELOVL fatty acid elongase 7; Very long chain 3-ketoacyl-CoA synthase 7; Very long chain 3-oxoacyl-CoA synthase 7; very-long-chain 3-oxoacyl-CoA synthase 7
Gene Aliases: ELOVL7
UniProt ID: (Human) A1L3X0
Entrez Gene ID: (Human) 79993
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.