Invitrogen
Manufacturer Code:PA513455
Catalog # PIPA513455
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 32-58 amino acids from the N-terminal region of human ELOVL6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-keto acyl-CoA synthase ELOVL6; Elongation of very long chain fatty acids protein 6; Elongation of very long chain fatty acids protein 6 hELO2 Fatty acyl-CoA elongase Long-chain fatty-acyl elongase LCE FACE ELOVL6; ELOVL FA elongase 6; ELOVL family member 6, elongation of long chain fatty acids (FEN1/Elo2, SUR4/Elo3-like, yeast); ELOVL fatty acid elongase 6; Fatty acid elongase 2; Fatty acyl-CoA elongase; hELO2; Long chain fatty acid elongase; long chain fatty acyl elongase; Long-chain fatty-acyl elongase; Myelin-associated SUR4 protein; Very long chain 3-ketoacyl-CoA synthase 6; Very long chain 3-oxoacyl-CoA synthase 6; very-long-chain 3-oxoacyl-CoA synthase 6
Gene Aliases: C77826; ELOVL6; FACE; FAE; LCE; Masr
UniProt ID: (Human) Q9H5J4, (Mouse) Q920L5
Entrez Gene ID: (Human) 79071, (Mouse) 170439
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.