Invitrogen
Manufacturer Code:PA5115176
Catalog # PIPA5115176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ELOVL5 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110059L23Rik 3-keto acyl-CoA synthase ELOVL5 AI747313 AU043003 dJ483K16.1 elongation of long chain fatty acids family member 5 elongation of very long chain fatty acids protein 5 elongation of very long chain fatty acids-like 5 ELOVL FA elongase 5 ELOVL family member 5 elongation of long chain fatty acids ELOVL family member 5 elongation of long chain fatty acids (FEN1/Elo2 SUR4/Elo3-like yeast) ELOVL family member 5 elongation of long chain fatty acids (yeast) ELOVL fatty acid elongase 5 ELOVL2 ELOVL5 fatty acid elongase 1 hELO1 homolog of yeast long chain polyunsaturated fatty acid elongation enzyme homolog of yeast long chain polyunsaturated fatty acid elongation enzyme 2 PRO0530 rELO1 SCA38 spinocerebellar ataxia 38 very long chain 3-ketoacyl-CoA synthase 5 Very long chain 3-oxoacyl-CoA synthase 5 very-long-chain 3-oxoacyl-CoA synthase 5; 3-keto acyl-CoA synthase ELOVL5; Elongation of very long chain fatty acids protein 5; ELOVL FA elongase 5; ELOVL family member 5, elongation of long chain fatty acids (FEN1/Elo2, SUR4/Elo3-like, yeast); ELOVL fatty acid elongase 5; Fatty acid elongase 1; hELO1; homolog of yeast long chain polyunsaturated fatty acid elongation enzyme; homolog of yeast long chain polyunsaturated fatty acid elongation enzyme 2; spinocerebellar ataxia 38; Very long chain 3-ketoacyl-CoA synthase 5; Very long chain 3-oxoacyl-CoA synthase 5; very-long-chain 3-oxoacyl-CoA synthase 5
Gene Aliases: 1110059L23Rik; AI747313; AU043003; dJ483K16.1; ELOVL2; ELOVL5; HELO1; PRO0530; SCA38
UniProt ID: (Human) Q9NYP7, (Mouse) Q8BHI7
Entrez Gene ID: (Human) 60481, (Mouse) 68801
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.