Invitrogen
Manufacturer Code:PA5101577
Catalog # PIPA5101577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ELOVL4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-keto acyl-CoA synthase ELOVL4; 3-keto acyl-CoA synthase ELOVL4 ADMD cancer/testis antigen 118 CT118 elongation of very long chain fatty acids (FEN1/Elo2 SUR4/Elo3 yeast)-like 4 elongation of very long chain fatty acids protein 4 elongation of very long chain fatty acids-like 4 ELOVL FA elongase 4 ELOVL fatty acid elongase 4 Elovl4 FLJ17667 FLJ92876 ISQMR SCA34 STGD2 STGD3 Very long chain 3-ketoacyl-CoA synthase 4 very long chain 3-oxoacyl-CoA synthase 4 very-long-chain 3-oxoacyl-CoA synthase 4; cancer/testis antigen 118; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 4; Elongation of very long chain fatty acids protein 4; ELOVL FA elongase 4; ELOVL fatty acid elongase 4; Very long chain 3-ketoacyl-CoA synthase 4; Very long chain 3-oxoacyl-CoA synthase 4; very-long-chain 3-oxoacyl-CoA synthase 4
Gene Aliases: ADMD; CT118; ELOVL4; ISQMR; SCA34; STGD2; STGD3
UniProt ID: (Human) Q9GZR5, (Mouse) Q9EQC4
Entrez Gene ID: (Human) 6785, (Mouse) 83603
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.