Novus Biologicals
Manufacturer Code:NBP250385AF594
Catalog # NP2038A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | V61 P2B3&asteriskD10 |
Immunogen | Ovalbumin-conjugated synthetic peptide - QPYNFELSK |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 3-keto acyl-CoA synthase ELOVL3; 3-keto acyl-CoA synthase ELOVL3 CIG-30 CIG30MGC21435 Cold-inducible glycoprotein of 30 kDa EC 2.3.1.n8 elongation of very long chain fatty acids (FEN1/Elo2 SUR4/Elo3 yeast)-like 3 elongation of very long chain fatty acids protein 3; Cold-inducible glycoprotein of 30 kDa; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 3; Elongation of very long chain fatty acids protein 3; ELOVL FA elongase 3; ELOVL fatty acid elongase 3; Very long chain 3-ketoacyl-CoA synthase 3; Very long chain 3-oxoacyl-CoA synthase 3; very-long-chain 3-oxoacyl-CoA synthase 3
Gene Aliases: CIG-30; CIG30; ELOVL3
UniProt ID: (Human) Q9HB03
Entrez Gene ID: (Human) 83401
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.