Invitrogen
Manufacturer Code:PA5103826
Catalog # PIPA5103826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ELOVL2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-keto acyl-CoA synthase ELOVL2; 3-keto acyl-CoA synthase Elovl2 AI317360 ELG3 elongation of very long chain fatty acids (FEN1/Elo2 SUR4/Elo3 yeast)-like 2 elongation of very long chain fatty acids protein 2 elongation of very long chain fatty acids-like 2 ELOVL FA elongase 2 ELOVL family member 2 ELOVL fatty acid elongase 2 ELOVL2 sequence similarity to Cig30 2 Ssc2 Very long chain 3-ketoacyl-CoA synthase 2 very long chain 3-oxoacyl-CoA synthase 2 very-long-chain 3-oxoacyl-CoA synthase 2; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 2; Elongation of very long chain fatty acids protein 2; ELOVL FA elongase 2; ELOVL fatty acid elongase 2; Very long chain 3-ketoacyl-CoA synthase 2; Very long chain 3-oxoacyl-CoA synthase 2; very-long-chain 3-oxoacyl-CoA synthase 2
Gene Aliases: ELG3; ELOVL2; SSC2
UniProt ID: (Human) Q9NXB9
Entrez Gene ID: (Human) 54898
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.