Invitrogen
Manufacturer Code:PA563320
Catalog # PIPA563320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ELOVL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-keto acyl-CoA synthase ELOVL1; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 1; Elongation of very long chain fatty acids protein 1; ELOVL FA elongase 1; ELOVL fatty acid elongase 1; SSC1; Very long chain 3-ketoacyl-CoA synthase 1; Very long chain 3-oxoacyl-CoA synthase 1; very-long-chain 3-oxoacyl-CoA synthase 1
Gene Aliases: CGI-88; ELOVL1; SSC1
UniProt ID: (Human) Q9BW60
Entrez Gene ID: (Human) 64834
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.