OriGene
Manufacturer Code:TA810828
Catalog # 50-168-3662
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 302-621 of human ELL produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C19orf17 ELL1 MEN PPP1R68; Eleven-nineteen lysine-rich leukemia protein; ELL gene (11-19 lysine-rich leukemia gene); elongation factor RNA polymerase II; protein phosphatase 1, regulatory subunit 68; RNA polymerase II elongation factor ELL
Gene Aliases: C19orf17; ELL; ELL1; MEN; PPP1R68
UniProt ID: (Human) P55199
Entrez Gene ID: (Human) 8178
Molecular Function: cell junction protein tight junction transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.