Invitrogen
Manufacturer Code:PA541513
Catalog # PIPA541513
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ELL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Eleven-nineteen lysine-rich leukemia protein; ELL gene (11-19 lysine-rich leukemia gene); elongation factor RNA polymerase II; protein phosphatase 1, regulatory subunit 68; RNA polymerase II elongation factor ELL; RNA POLYMERASE II ELONGATION FACTOR ELL (ELEVEN-NINETEEN LYSINE-RICH LEUKEMIA PROTEIN)
Gene Aliases: C19orf17; ELL; ELL1; MEN; PPP1R68
UniProt ID: (Human) P55199, (Mouse) O08856
Entrez Gene ID: (Dog) 484820, (Human) 8178, (Bovine) 506578, (Guinea pig) 100718569, (Mouse) 13716, (Rat) 306347
Molecular Function:
cell junction protein
tight junction
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.