OriGene
Manufacturer Code:TA810143
Catalog # 50-168-3219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E11 |
Immunogen | Human recombit protein fragment corresponding to amino acids 83-345 of human ELF4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E74-like factor 4; E74-like factor 4 (ets domain transcription factor); ELF4; ELFR MEF; ETS-related transcription factor Elf-4; Myeloid Elf-1-like factor
Gene Aliases: ELF4; ELFR; MEF
UniProt ID: (Human) Q99607
Entrez Gene ID: (Human) 2000
Molecular Function: nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.