Invitrogen
Manufacturer Code:PA577157
Catalog # PIPA577157
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human ELF4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E74-like factor 4; E74-like factor 4 (ets domain transcription factor); ELF4; ETS-related transcription factor Elf-4; ETS-related transcription factor Elf-4 E74-like factor 4 Myeloid Elf-1-like factor ELF4 ELFR MEF; Myeloid Elf-1-like factor
Gene Aliases: ELF4; ELFR; MEF
UniProt ID: (Human) Q99607
Entrez Gene ID: (Human) 2000
Molecular Function: nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.