Invitrogen
Manufacturer Code:PA575506
Catalog # PIPA575506
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 219-264 of Human ELF4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E74-like factor 4; E74-like factor 4 (ets domain transcription factor); E74-like factor 4; myeloid elf-1-like factor; myeloid elf-1 like factor; ELF4; ETS-related transcription factor Elf-4; ETS-related transcription factor Elf-4 E74-like factor 4 Myeloid Elf-1-like factor ELF4 ELFR MEF; myeloid elf-1 like factor; Myeloid Elf-1-like factor
Gene Aliases: AV314029; BC042423; ELF4; ELFR; ENSMUSG00000053512; Gm9907; MEF; RGD1560743
UniProt ID: (Human) Q99607, (Mouse) Q9Z2U4
Entrez Gene ID: (Human) 2000, (Mouse) 56501, (Rat) 302811
Molecular Function: nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.