Invitrogen
Manufacturer Code:MA527425
Catalog # PIMA527425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D3 |
Immunogen | Full length human recombit protein of ELF1 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E74 like factor 1 E74-like factor 1 ELF 1 ELF1 ELF1 Ets domain transcription factor ETS related transcription factor Elf 1 ETS related transcription factor Elf1 ets-related transcription factor ETS-related transcription factor Elf-1 mElf 1; E74-like factor 1; E74-like factor 1 (ets domain transcription factor); ETS-related transcription factor Elf-1
Gene Aliases: ELF1
UniProt ID: (Human) P32519
Entrez Gene ID: (Human) 1997
Molecular Function:
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.