Invitrogen
Manufacturer Code:PA579200
Catalog # PIPA579200
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human E74 like factor 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E74-like factor 1; E74-like factor 1 (ets domain transcription factor); ETS-related transcription factor Elf-1; ETS-related transcription factor Elf-1 E74-like factor 1 ELF1
Gene Aliases: Elf-1; ELF1; mElf-1; p70; Sts1
UniProt ID: (Human) P32519, (Mouse) Q60775
Entrez Gene ID: (Human) 1997, (Mouse) 13709, (Rat) 85424
Molecular Function:
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.