Invitrogen
Manufacturer Code:PA554016
Catalog # PIPA554016
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ELAC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: elaC homolog 2; ElaC homolog protein 2; elaC-like protein 2; ELC2 HPC2 RNase Z 2 elaC homolog protein 2 heredity prostate cancer protein 2 putative prostate cancer susceptibility protein HPC2/ELAC2 ribonuclease Z 2 tRNA 3 endonuclease 2 tRNase Z (long form) tRNase Z 2 zinc phosphodiesterase ELAC protein 2; Heredity prostate cancer protein 2; putative prostate cancer susceptibility protein HPC2/ELAC2; Ribonuclease Z 2; RNase Z 2; tRNA 3 endonuclease 2; tRNase Z (long form); tRNase Z 2; Zinc phosphodiesterase ELAC protein 2
Gene Aliases: COXPD17; ELAC2; ELC2; HPC2
UniProt ID: (Human) Q9BQ52
Entrez Gene ID: (Human) 60528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.