Novus Biologicals
Manufacturer Code:NBP297772IR
Catalog # NB103399
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human ELAC2 fragment. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.26.11 elaC homolog 2 (E. coli) ElaC homolog protein 2 ELC2 FLJ10530 FLJ36693 FLJ42848 Heredity prostate cancer protein 2 HPC2elaC (E. coli) homolog 2 putative prostate cancer susceptibility protein HPC2/ELAC2 Ribonuclease Z 2 RNase Z 2 tRNA 3 endonuclease 2 tRNase Z (long form) tRNase Z 2 zinc phosphodiesterase ELAC protein 2; elaC homolog 2; ElaC homolog protein 2; elaC-like protein 2; Heredity prostate cancer protein 2; putative prostate cancer susceptibility protein HPC2/ELAC2; Ribonuclease Z 2; RNase Z 2; tRNA 3 endonuclease 2; tRNase Z (long form); tRNase Z 2; Zinc phosphodiesterase ELAC protein 2
Gene Aliases: COXPD17; ELAC2; ELC2; HPC2
UniProt ID: (Human) Q9BQ52
Entrez Gene ID: (Human) 60528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.