Proteintech
Manufacturer Code:100711AP150UL
Catalog # 50-172-6000
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ELAC2 Fusion Protein Ag0105 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: elaC homolog 2; elaC homolog 2 (E. coli) ElaC homolog protein 2 ELAC2 ELC2 HPC2 Ribonuclease Z 2 RNase Z 2 tRNA 3 endonuclease 2 tRNase Z 2; ElaC homolog protein 2; elaC-like protein 2; Heredity prostate cancer protein 2; putative prostate cancer susceptibility protein HPC2/ELAC2; Ribonuclease Z 2; RNase Z 2; tRNA 3 endonuclease 2; tRNase Z (long form); tRNase Z 2; tRNase Z(L); Zinc phosphodiesterase ELAC protein 2
Gene Aliases: 1110017O07Rik; COXPD17; D11Wsu80e; ELAC2; ELC2; HPC2
UniProt ID: (Human) Q9BQ52, (Mouse) Q80Y81, (Rat) Q8CGS5
Entrez Gene ID: (Human) 60528, (Mouse) 68626, (Rat) 282826
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.