Abnova
Manufacturer Code:H00060528K
Catalog # 89-204-034
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ELAC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ELAC2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: elaC homolog 2; ElaC homolog protein 2; elaC-like protein 2; ELC2 FLJ10530 FLJ36693 FLJ42848 HPC2; Heredity prostate cancer protein 2; putative prostate cancer susceptibility protein HPC2/ELAC2; Ribonuclease Z 2; RNase Z 2; tRNA 3 endonuclease 2; tRNase Z (long form); tRNase Z 2; Zinc phosphodiesterase ELAC protein 2
Gene Aliases: COXPD17; ELAC2; ELC2; HPC2
UniProt ID: (Human) Q9BQ52
Entrez Gene ID: (Human) 60528
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.