Invitrogen
Manufacturer Code:PA559332
Catalog # PIPA559332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ELAC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D29 RNase Z 1 deleted in Ma29 elaC homolog protein 1 ribonuclease Z 1 tRNA 3 endonuclease 1 tRNA 3&prime processing endoribonuclease tRNA Z (short form) tRNase Z 1 tRNase ZS zinc phosphodiesterase ELAC protein 1; Deleted in Ma29; elaC homolog 1; ElaC homolog protein 1; Ribonuclease Z 1; RNase Z 1; RNaseZ(S); tRNA 3 endonuclease 1; tRNA 3' processing endoribonuclease; tRNA Z (short form); tRNase Z 1; tRNase ZS; Zinc phosphodiesterase ELAC protein 1
Gene Aliases: 2610018O07Rik; 8430417G19Rik; D29; ELAC1
UniProt ID: (Human) Q9H777, (Mouse) Q8VEB6
Entrez Gene ID: (Human) 55520, (Rat) 307604, (Mouse) 114615
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.