Proteintech
Manufacturer Code:266391AP150UL
Catalog # 50-173-5410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ELAC1 Fusion Protein Ag23939 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D29 Deleted in Ma29 elaC homolog 1 (E. coli) ElaC homolog protein 1 ELAC1 Ribonuclease Z 1 RNase Z 1 tRNA 3 endonuclease 1 tRNase Z 1; Deleted in Ma29; elaC homolog 1; ElaC homolog protein 1; Ribonuclease Z 1; RNase Z 1; RNaseZ(S); tRNA 3 endonuclease 1; tRNA 3' processing endoribonuclease; tRNA Z (short form); tRNase Z 1; tRNase ZS; Zinc phosphodiesterase ELAC protein 1
Gene Aliases: 2610018O07Rik; 8430417G19Rik; D29; ELAC1
UniProt ID: (Human) Q9H777, (Mouse) Q8VEB6
Entrez Gene ID: (Human) 55520, (Mouse) 114615
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.