OriGene
Manufacturer Code:TA505147
Catalog # 50-167-5686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6F7 |
Immunogen | Full length human recombit protein of human EIF5A2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: eIF-5A-2; EIF-5A2 eIF5AII; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 2; Eukaryotic translation initiation factor 5A-2
Gene Aliases: EIF-5A2; EIF5A2; eIF5AII
UniProt ID: (Human) Q9GZV4
Entrez Gene ID: (Human) 56648
Molecular Function: RNA binding protein nucleic acid binding translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.