Invitrogen
Manufacturer Code:PA530770
Catalog # PIPA530770
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 153 of Human EIF5A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-5A-2; EIF-5A2 eIF5AII eIF-5A-2 eukaryotic initiation factor 5A eukaryotic translation initiation factor 5A-2; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 2; Eukaryotic translation initiation factor 5A-2
Gene Aliases: EIF-5A2; EIF5A2; eIF5AII
UniProt ID: (Human) Q9GZV4
Entrez Gene ID: (Human) 56648
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.