Proteintech
Manufacturer Code:110121AP150UL
Catalog # 50-172-6794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EIF4H Fusion Protein Ag1468 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF 4H EIF4H KIAA0038 WBSCR1 WSCR1; eIF-4H; Eukaryotic translation initiation factor 4H; Williams-Beuren syndrome chromosomal region 1 protein; Williams-Beuren syndrome chromosome region 1
Gene Aliases: eIF-4H; EIF4H; KIAA0038; WBSCR1; WSCR1
UniProt ID: (Human) Q15056
Entrez Gene ID: (Human) 7458
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.