Novus Biologicals
Manufacturer Code:NBP271468JF646
Catalog # N271468J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F11 |
Immunogen | Full length recombinant protein of human EIF4E2 (NP_004837) produced in HEK293T cells. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 4EHP 4E-LP eIF4E type 2 eIF-4E type 2 EIF4EL3 eIF4E-like cap-binding protein eIF4E-like protein 4E-LP eukaryotic translation initiation factor 4E family member 2 Eukaryotic translation initiation factor 4E homologous protein eukaryotic translation initiation factor 4E member 2 eukaryotic translation initiation factor 4E type 2 Eukaryotic translation initiation factor 4E-like 34E-LP IF4e mRNA cap-binding protein 4EHP mRNA cap-binding protein type 3; eIF-4E type 2; eIF4E-like cap-binding protein; eIF4E-like protein 4E-LP; Eukaryotic translation initiation factor 4E homologous protein; Eukaryotic translation initiation factor 4E type 2; Eukaryotic translation initiation factor 4E-like 3; h4EHP; mRNA cap-binding protein 4EHP; mRNA cap-binding protein type 3
Gene Aliases: 4E-LP; 4EHP; EIF4E2; EIF4EL3; IF4e
UniProt ID: (Human) O60573
Entrez Gene ID: (Human) 9470
Molecular Function: RNA binding protein nucleic acid binding translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.