Proteintech
Manufacturer Code:122271AP150UL
Catalog # 50-172-7812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EIF4E2 Fusion Protein Ag2864 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4E LP 4EHP eIF 4E type 2 eIF4E like protein 4E LP eIF4E type 2 EIF4E2 EIF4EL3 IF4e mRNA cap binding protein 4EHP; eIF-4E type 2; eIF4E type 2; eIF4E-2; eIF4E-like cap-binding protein; eIF4E-like protein 4E-LP; Eukaryotic translation initiation factor 4E homologous protein; eukaryotic translation initiation factor 4E like 3; eukaryotic translation initiation factor 4E member 2; Eukaryotic translation initiation factor 4E type 2; Eukaryotic translation initiation factor 4E-like 3; h4EHP; mRNA cap-binding protein 4EHP; mRNA cap-binding protein type 2; mRNA cap-binding protein type 3
Gene Aliases: 2700069E09Rik; 4E-LP; 4EHP; AI036339; AV129531; D0H0S6743E; EIF4E2; EIF4EL3; IF4e
UniProt ID: (Human) O60573, (Mouse) Q8BMB3
Entrez Gene ID: (Human) 9470, (Rat) 363275, (Mouse) 26987
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.