Abnova
Manufacturer Code:PAB12256
Catalog # 89-115-680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to residues surrounding S422 of human EIF4B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of EIF4B.
For Research Use Only
Protein Aliases: EIF-4B; Eukaryotic translation initiation factor 4B; PRO1843
Gene Aliases: EIF-4B; EIF4B; PRO1843
UniProt ID: (Human) P23588
Entrez Gene ID: (Human) 1975
Molecular Function: RNA binding protein nucleic acid binding translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.