Invitrogen
Manufacturer Code:PA5116950
Catalog # PIPA5116950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human EIF4A1/EIF4A2/EIF4A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase DDX48; ATP-dependent RNA helicase eIF4A-1; ATP-dependent RNA helicase eIF4A-2; ATP-dependent RNA helicase eIF4A-3; DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; DEAD box protein 48; eIF-4A-I; eIF-4A-II; eIF-4A-III; eIF4A-I; eIF4A-II; eIF4A-III; Eukaryotic initiation factor 4A-I; Eukaryotic initiation factor 4A-II; Eukaryotic initiation factor 4A-III; Eukaryotic initiation factor 4A-III, N-terminally processed; Eukaryotic initiation factor 4A-like NUK-34; eukaryotic initiation factor 4AI; Eukaryotic translation initiation factor 4A isoform 3; eukaryotic translation initiation factor 4A, isoform 1; eukaryotic translation initiation factor 4A, isoform 2; eukaryotic translation initiation factor 4A, isoform 3; hNMP 265; initiation factor eIF-4A long form; NMP 265; Nuclear matrix protein 265; protein synthesis initiation factor 4AII
Gene Aliases: 2400003O03Rik; 4833432N07Rik; BM-010; DDX2A; DDX2B; DDX48; EIF-4A; eIF-4A-I; eIF-4A-II; Eif4; EIF4A; eIF4A-I; eIF4A-II; eIF4A-III; EIF4A1; EIF4A2; EIF4A3; eIF4AIII; EIF4F; KIAA0111; mKIAA0111; MUK34; NMP265; NUK34; RCPS
UniProt ID: (Human) P60842, (Human) Q14240, (Human) P38919, (Mouse) P60843, (Mouse) P10630, (Rat) Q5RKI1, (Mouse) Q91VC3, (Rat) Q3B8Q2
Entrez Gene ID: (Human) 1973, (Human) 1974, (Human) 9775, (Mouse) 13681, (Rat) 287436, (Mouse) 13682, (Rat) 303831, (Mouse) 192170, (Rat) 688288
Molecular Function: RNA binding protein RNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.