Proteintech
Manufacturer Code:104391AP150UL
Catalog # 50-172-6309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EIF3J Fusion Protein Ag0678 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF 3 alpha eIF3 alpha eIF3 p35 EIF3J EIF3S1; eIF-3-alpha; eIF-3-alpha {ECO:0000255|HAMAP-Rule:MF_03009}; eIF-3-alpha-A; eIF-3-alpha-B; eIF3 p35; eIF3 p35 {ECO:0000255|HAMAP-Rule:MF_03009}; EIF3J; eIF3j {ECO:0000255|HAMAP-Rule:MF_03009}; eIF3j-A; eIF3j-B; Eukaryotic translation initiation factor 3 subunit 1; eukaryotic translation initiation factor 3 subunit 1 {ECO:0000255|HAMAP-Rule:MF_03009}; Eukaryotic translation initiation factor 3 subunit 1-A; eukaryotic translation initiation factor 3 subunit 1-B; Eukaryotic translation initiation factor 3 subunit J; eukaryotic translation initiation factor 3 subunit J {ECO:0000255|HAMAP-Rule:MF_03009}; Eukaryotic translation initiation factor 3 subunit J-A; eukaryotic translation initiation factor 3, subunit 1 (alpha, 35kD); eukaryotic translation initiation factor 3, subunit 1 alpha; eukaryotic translation initiation factor 3, subunit J
Gene Aliases: 1810016I04Rik; 2700079K05Rik; AA409117; AA409446; C78575; eIF-3-alpha-A; eIF-3-alpha-B; eIF3-alpha; eIF3-p35; EIF3J; Eif3j-1; Eif3j-2; eIF3j-A; eIF3j-B; Eif3j1; Eif3j2; EIF3S1; Eif3s1-1; PRO0391
UniProt ID: (Human) O75822, (Rat) A0JPM9, (Mouse) Q3UGC7
Entrez Gene ID: (Human) 8669, (Rat) 691947, (Mouse) 78655
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.