Proteintech
Manufacturer Code:660241IG150UL
Catalog # 50-173-6466
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C5A3 |
Immunogen | EIF3D Fusion Protein Ag18092 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: eIF 3 zeta eIF3 p66 eIF3 zeta EIF3D EIF3S7; eIF-3-zeta; eIF-3-zeta {ECO:0000255|HAMAP-Rule:MF_03003}; eIF3 p66; EIF3D; eIF3d {ECO:0000255|HAMAP-Rule:MF_03003}; Eukaryotic translation initiation factor 3 subunit 7; eukaryotic translation initiation factor 3 subunit 7 {ECO:0000255|HAMAP-Rule:MF_03003}; Eukaryotic translation initiation factor 3 subunit D; eukaryotic translation initiation factor 3 subunit D {ECO:0000255|HAMAP-Rule:MF_03003}; eukaryotic translation initiation factor 3, subunit 7 (zeta); eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa; eukaryotic translation initiation factor 3, subunit D; translation initiation factor eIF3 p66; translation initiation factor eIF3 p66 subunit
Gene Aliases: 66/67kDa; AA407891; eIF3-p66; eIF3-zeta; EIF3D; eIF3p66; EIF3S7
UniProt ID: (Human) O15371, (Rat) Q6AYK8, (Mouse) O70194
Entrez Gene ID: (Human) 8664, (Rat) 362952, (Mouse) 55944
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.