OriGene
Manufacturer Code:TA501313
Catalog # 50-167-2459
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3H4 |
Immunogen | Full length human recombit protein of human EIF2S1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EIF-2 EIF-2A EIF-2alpha EIF2 EIF2A; eIF-2-alpha; EIF-2A; EIF-2alpha; Eukaryotic translation initiation factor 2 subunit 1; Eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa
Gene Aliases: EIF-2; EIF-2A; EIF-2alpha; EIF2; EIF2A; EIF2S1
UniProt ID: (Human) P05198, (Rat) P68101
Entrez Gene ID: (Dog) 480361, (Human) 1965, (Rat) 54318
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.