Proteintech
Manufacturer Code:196921AP150UL
Catalog # 50-173-2490
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGO3 Argonaute 3 Argonaute3 eIF 2C 3 eIF2C 3 EIF2C3 hAgo3 Protein argonaute 3; argonaute 3; Argonaute RISC catalytic component 3; Argonaute3; eIF-2C 3; eIF2C 3; Eukaryotic translation initiation factor 2C 3; eukaryotic translation initiation factor 2C, 3; hAgo3; mAgo3; Piwi/Argonaute family protein meIF2C3; Protein argonaute-3
Gene Aliases: AGO3; AW048688; C130014L07Rik; EIF2C3
UniProt ID: (Human) Q9H9G7, (Mouse) Q8CJF9
Entrez Gene ID: (Human) 192669, (Mouse) 214150, (Rat) 313593
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.