Proteintech
Manufacturer Code:196901AP150UL
Catalog # 50-173-2489
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGO1 Argonaute1 eIF 2C 1 EIF2C eIF2C 1 EIF2C1 GERP95 hAgo1 Protein argonaute 1 Q99; argonaute 1; Argonaute RISC catalytic component 1; Argonaute1; eIF-2C 1; eIF2C 1; Eukaryotic translation initiation factor 2C 1; eukaryotic translation initiation factor 2C, 1; Golgi Endoplasmic Reticulum protein 95 kDa; mAgo1; Piwi/Argonaute family protein meIF2C1; Protein argonaute-1; Putative RNA-binding protein Q99
Gene Aliases: AGO1; EIF2C; EIF2C1; GERP95; hAgo1; Q99
UniProt ID: (Human) Q9UL18, (Mouse) Q8CJG1
Entrez Gene ID: (Human) 26523, (Mouse) 236511, (Rat) 313594
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.