Abnova
Manufacturer Code:H00026523K
Catalog # 89-200-592
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human EIF2C1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human EIF2C1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: AGO1 DKFZp686M13167 EIF2C GERP95 Q99; argonaute 1; Argonaute RISC catalytic component 1; Argonaute1; eIF-2C 1; eIF2C 1; Eukaryotic translation initiation factor 2C 1; eukaryotic translation initiation factor 2C, 1; Golgi Endoplasmic Reticulum protein 95 kDa; Protein argonaute-1; Putative RNA-binding protein Q99
Gene Aliases: AGO1; EIF2C; EIF2C1; GERP95; hAgo1; Q99
UniProt ID: (Human) Q9UL18
Entrez Gene ID: (Human) 26523
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.