Proteintech
Manufacturer Code:113321AP150UL
Catalog # 50-555-135
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EIF2B4 Fusion Protein Ag1875 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EIF 2B EIF2B EIF2B4 EIF2BD EIF2Bdelta eIF2Bdelta; eIF-2B GDP-GTP exchange factor subunit delta; eukaryotic translation initiation factor 2B subunit 4 delta; eukaryotic translation initiation factor 2B, subunit 4 delta, 67kDa; translation initiation factor eIF-2b delta subunit; Translation initiation factor eIF-2B subunit delta
Gene Aliases: EIF-2B; EIF2B; EIF2B4; EIF2BD; EIF2Bdelta; Jgr1a
UniProt ID: (Human) Q9UI10, (Rat) Q63186, (Mouse) Q61749
Entrez Gene ID: (Human) 8890, (Rat) 117019, (Mouse) 13667
Molecular Function:
RNA binding protein
isomerase
nucleic acid binding
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.