Novus Biologicals
Manufacturer Code:NB10075461
Catalog # NB10075461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody was a sequence from 1a.a. to 210a.a. from EIF2B2(a 351a.a. long protein) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: eIF-2B GDP-GTP exchange factor subunit beta; EIF2B eIF-2B GDP-GTP exchange factor subunit beta EIF2BB EIF-2Bbeta eukaryotic translation initiation factor 2B subunit 2 (beta 39kD) eukaryotic translation initiation factor 2B subunit 2 beta 39kDa S20I15 S20III15 translation initiation factor eIF-2B subunit beta; eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa; S20I15; S20III15; Translation initiation factor eIF-2B subunit beta
Gene Aliases: EIF-2Bbeta; EIF2B; EIF2B2; EIF2BB
UniProt ID: (Human) P49770
Entrez Gene ID: (Human) 8892
Molecular Function: RNA binding protein isomerase nucleic acid binding translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.