Abnova
Manufacturer Code:H00008892K
Catalog # 89-209-719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human EIF2B2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human EIF2B2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: eIF-2B GDP-GTP exchange factor subunit beta; EIF-2Bbeta EIF2B; eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa; S20I15; S20III15; Translation initiation factor eIF-2B subunit beta
Gene Aliases: EIF-2Bbeta; EIF2B; EIF2B2; EIF2BB
UniProt ID: (Human) P49770
Entrez Gene ID: (Human) 8892
Molecular Function: RNA binding protein isomerase nucleic acid binding translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.