Abnova
Manufacturer Code:PAB29217
Catalog # 89-130-651
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T982 of human EIF2AK3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EIF2AK3.
For Research Use Only
Protein Aliases: DKFZp781H1925; Eukaryotic translation initiation factor 2-alpha kinase 3; HRI; HsPEK; Pancreatic eIF2-alpha kinase; PEK; PERK; PRKR-like endoplasmic reticulum kinase; WRS
Gene Aliases: EIF2AK3; PEK; PERK; WRS
UniProt ID: (Human) Q9NZJ5
Entrez Gene ID: (Human) 9451
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.