Novus Biologicals
Manufacturer Code:NBP2829840.1ML
Catalog # NB030913
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of EID2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CREBBP/EP300 inhibitor 2; CREBBP/EP300 inhibitor 2E1A like inhibitor of differentiation 2 CREBBP/EP300 inhibitory protein 2 CRI2EP300-interacting inhibitor of differentiation 2 EID-2EID-1-like inhibitor of differentiation 2 EP300 interacting inhibitor of differentiation 2 MGC20452; CREBBP/EP300 inhibitory protein 2; E1A like inhibitor of differentiation 2; EID-1-like inhibitor of differentiation 2; EID-2; EID2; EP300-interacting inhibitor of differentiation 2
Gene Aliases: CRI2; EID-2; EID2
UniProt ID: (Human) Q8N6I1
Entrez Gene ID: (Human) 163126
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.