Abnova
Manufacturer Code:PAB24418
Catalog # 89-126-131
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human EID2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant EID2.
For Research Use Only
Protein Aliases: CREBBP/EP300 inhibitor 2; CREBBP/EP300 inhibitory protein 2; CRI2; E1A like inhibitor of differentiation 2; EID-1-like inhibitor of differentiation 2; EID-2; EID2; EP300-interacting inhibitor of differentiation 2; MGC20452
Gene Aliases: CRI2; EID-2; EID2
UniProt ID: (Human) Q8N6I1
Entrez Gene ID: (Human) 163126
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.