Novus Biologicals
Manufacturer Code:NBP297540AF700
Catalog # NB044669
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human EID1 fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 21 kDa pRb-associated protein; 21 kDa pRb-associated protein C15orf3PTD014 CREBBP/EP300 inhibitor 1 CREBBP/EP300 inhibitory protein 1MGC138884 CRI1EP300-interacting inhibitor of differentiation 1 E1A-like inhibitor of differentiation 1 EID-1MGC138883 EP300 interacting inhibitor of differentiation 1 IRO45620 NB4 apoptosis related protein Rb- and p300-binding protein EID-1 RBP21 retinoblastoma protein-associated protein; CREBBP/EP300 inhibitor 1; CREBBP/EP300 inhibitory protein 1; E1A-like inhibitor of differentiation 1; EID-1; EID1; EP300-interacting inhibitor of differentiation 1; NB4 apoptosis related protein; Rb- and p300-binding protein EID-1; retinoblastoma protein-associated protein
Gene Aliases: C15orf3; CRI1; EID-1; EID1; IRO45620; PNAS-22; PTD014; RBP21
UniProt ID: (Human) Q9Y6B2
Entrez Gene ID: (Human) 23741
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.