Proteintech
Manufacturer Code:117341AP150UL
Catalog # 50-172-7413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | EID1 Fusion Protein Ag2336 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 21 kDa pRb associated protein C15orf3 CRI1 EID 1 EID1 IRO45620 PNAS 22 PTD014 RBP21; 21 kDa pRb-associated protein; CREBBP/EP300 inhibitor 1; CREBBP/EP300 inhibitory protein 1; E1A-like inhibitor of differentiation 1; EID-1; EID1; EP300-interacting inhibitor of differentiation 1; NB4 apoptosis related protein; Rb- and p300-binding protein EID-1; retinoblastoma protein-associated protein
Gene Aliases: 2610002K20Rik; C15orf3; CRI1; EID-1; EID1; IRO45620; ORF12; PNAS-22; PTD014; RBP21
UniProt ID: (Human) Q9Y6B2, (Mouse) Q9DCR4
Entrez Gene ID: (Human) 23741, (Mouse) 58521
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.