Invitrogen
Manufacturer Code:PA5102406
Catalog # PIPA5102406
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human EHMT2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ankyrin repeat-containing protein; ankyrin repeat-containing protein Bat8 C6orf30 D17Ertd710e DKFZp686H08213 Ehmt2 euchromatic histone lysine methyltransferase 2 euchromatic histone lysine N-methyltransferase 2 euchromatic histone-lysine N-methyltransferase 2 FLJ35547 G9A G9A histone methyltransferase GAT8 H3 ly H3-K9-Histone-Methyl-Transferase H3-K9-HMTase 3 Histone H3-K9 methyltransferase 3 Histone-lysine N-methyltransferase EHMT2 histone-lysine N-methyltransferase H3 lysine-9 specific 3 HLA-B associated transcript 8 HLA-B associated transcript 8 rat orthologue HLA-B-associated transcript 8 KMT1C Lysine N-methyltransferase 1C NG36 NG36/G9a Protein G9a; Euchromatic histone-lysine N-methyltransferase 2; G9A histone methyltransferase; H3-K9-HMTase 3; Histone H3-K9 methyltransferase 3; Histone-lysine N-methyltransferase EHMT2; histone-lysine N-methyltransferase, H3 lysine-9 specific 3; HLA-B associated transcript 8; HLA-B associated transcript 8, rat orthologue; HLA-B-associated transcript 8; Lysine N-methyltransferase 1C; Protein G9a
Gene Aliases: BAT8; C6orf30; D17Ertd710e; EHMT2; G9A; GAT8; KMT1C; NG36
UniProt ID: (Human) Q96KQ7, (Mouse) Q9Z148
Entrez Gene ID: (Human) 10919, (Mouse) 110147, (Rat) 361798
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.